Water penetration tester, acc. to Karsten
Catalog # SIMELI7500
Supplier: Industrial Physics
Specifications
- Description:Water penetration test kit, acc. to Karsten
- Environmentally Preferable:
- Pk:1
Specifications
About this item
This Karsten tube penetration test is a simple test for measuring the degree or water penetration into building materials such as concrete, stone and plaster. The test kit consists of a glass tube filled with water, bonded to the test material with plastiline. Water pressure is then exerted on the surface. A graduated scale indicates, over time, the amount or water penetrated into the surface.
- Tube material: Glass
- Suitable for vertical and horizontal surfaces
Ordering information: The TQC Karsten Tube Penetration Test contains 3 tubes for horizontal surfaces, 3 tubes for vertical surfaces (or at choice any other combination of total 6 tubes), a water bottle, marker and putty to place the tubes leak-tight on the surface. Additional vertical or horizontal tubes are available on request.