Microscope slides, Snowcoat®
Microscope slides, Snowcoat®, White, 26×76 mm
LEIH3808100GEP
These slides are designed for optimisal use with xylene-resistant pens or pencils.
- Ground edges
- Six colours available
- Easy colour-coded identification
- Colour:White
- Description:Microscope slides, Snowcoat®
- L×W:26×76 mm
- Seq:01