Genomic DNA isolation and purification kits, OmniTemplate™
786-013
:
- Cdt:100 Tests
- Method / Format:Extraction for direct PCR
- Prep size:Scalable
- Refine target molecule:Total DNA
- Sample type:Cultured cellsHuman/animal cells and tissues
- Target molecule:DNA
- Désignation:OmniTemplate™
- Désignation:OmniTemplate™
OmniTemplate™ is specifically designed for the rapid isolation of a DNA template for PCR analysis from mammalian tissue samples, blood and cell cultures. This single tube method produces a high concentration, ready-to-use supply of DNA template for microanalysis, genotyping and a multitude of other applications.
- Single tube genomic DNA isolation
- Compatible with PCR and other downstream applications
- Suitable for blood, cells, and animal tissue samples
- No toxic chemicals, no phenol, no hazardous waste