Orders LinkContactUsLinkComponent
Forceps, Axal™
Forceps, Axal™
Supplier:   OUTILS RUBIS
Forceps, Axal™
Supplier:  OUTILS RUBIS
Certificates
Forceps, Axal™
Forceps, Axal™
15 Options Available Below
Product Details & Documents
Cobalt-chromium-molybdenum alloy. These forceps with very high Vicker hardness offer a range of physical, chemical and technological properties that make them appropriate for a wide variety of applications. Exceptional good tolerance on human tissues.
  • High yield strength alloy
  • 100% antimagnetic, rust proof and acid resistant
  • Highly resistant to corrosion, stress corrosion cracking and hydrogen embattlement
  • Temperature resistance of 500° C
  • Good fatigue and tensile strength guarantee durability
Perfect forceps for surgical appliances and laboratories.
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • OUTI0-AXAL
    Straight, fine tips
    Straight, fine tips
    0 Axal™
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 105,00 €
    105,00 €
    1 * 1 ST
       105,00 €
  • OUTI1-AXAL
    Straight, fine tips
    Straight, fine tips
    1 Axal™
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 102,00 €
    102,00 €
    1 * 1 ST
       102,00 €
  • OUTI2A-AXAL
    Straight, blunt tips
    Straight, blunt tips
    2A Axal™
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 101,00 €
    101,00 €
    1 * 1 ST
       101,00 €
  • OUTI2-AXAL
    Straight, fine tips, tower tip
    Straight, fine tips, tower tip
    2 Axal™
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 105,00 €
    105,00 €
    1 * 1 ST
       105,00 €
  • OUTI3-AXAL
    Straight, fine tips, edges on the outside rounded off
    Straight, fine tips, edges on the outside rounded off
    3 Axal™
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 105,00 €
    105,00 €
    1 * 1 ST
       105,00 €
  • OUTI3C-AXAL
    Straight, fine tips, edges on the outside rounded off
    Straight, fine tips, edges on the outside rounded off
    3C Axal™
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €
    1 * 1 ST
       104,00 €
  • OUTI4AB-AXAL
    Straight, fine tips, hook-shaped
    Straight, fine tips, hook-shaped
    4AB Axal™
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 110,00 €
    110,00 €
    1 * 1 ST
       110,00 €
  • OUTI5A-AXAL
    Bent, fine tips
    Bent, fine tips
    5A Axal™
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 113,00 €
    113,00 €
    1 * 1 ST
       113,00 €
  • OUTI5-AXAL
    Straight, fine tips, tower tip
    Straight, fine tips, tower tip
    5 Axal™
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 117,00 €
    117,00 €
    1 * 1 ST
       117,00 €
  • OUTI7-AXAL
    Straight, fine tips, 40° hook-shaped, edges on the outside rounded off
    Straight, fine tips, 40° hook-shaped, edges on the outside rounded off
    7 Axal™
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €
  • OUTIF-AXAL
    Straight, square tips, tower tip
    Straight, square tips, tower tip
    F Axal™
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 101,00 €
    101,00 €
    1 * 1 ST
       101,00 €
  • OUTISS-AXAL
    Straight, slim, ultra fine tips
    Straight, slim, ultra fine tips
    SS Axal™
    135 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 110,00 €
    110,00 €
    1 * 1 ST
       110,00 €
  • OUTI0C-11-AXAL
    Straight, fine tips
    Straight, fine tips
    0C 11 Axal™
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 102,00 €
    102,00 €
    1 * 1 ST
       102,00 €
  • OUTI4-AXAL
    Straight, fine tips, tower tip
    Straight, fine tips, tower tip
    4 Axal™
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €
  • OUTI7A-AXAL
    Straight, fine tips, 40° hook-shaped
    Straight, fine tips, 40° hook-shaped
    7A Axal™
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €