Order Entry
Belgium
ContactUsLinkComponent
Genomic DNA isolation from Yeasts, OmniPrep™ for Yeast kit
Genomic DNA isolation from Yeasts, OmniPrep™ for Yeast kit
Catalog # 786-400
Supplier:  G-Biosciences
CAS Number:  
undefined
Genomic DNA isolation from Yeasts, OmniPrep™ for Yeast kit
Catalog # 786-400
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Precipitation
    Organic extraction
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Yeast/Fungi
  • Target molecule:
    DNA
  • Description:
    OmniPrep™ for Yeast Kit, for 300 ml culture
  • Description:
    OmniPrep™ for Yeast Kit
  • Yield:
    3 - 6 μg/ml yeast culture

Specifications

About this item

OmniPrep™ for Yeast Kit isolates high quality genomic DNA from yeast. When used according to the recommended protocols, this kit purifies DNA from 300 ml yeast culture.

  • High Yield, ~100 kb genomic DNA
  • A₂₆₀/A₂₈₀ between 1,8 and 2,0
  • Simple two tube method, 20 to 40 minutes
  • Supplied with Yeast Suspension Buffer and LongLife™ Zymolyase® for efficient DNA release
  • No toxic chemicals, no phenol, no hazardous waste