Order Entry
Belgium
ContactUsLinkComponent
Genomic DNA isolation from plants, OmniPrep™
Genomic DNA isolation from plants, OmniPrep™
Catalog # 786-397
Supplier:  G-Biosciences
CAS Number:  
undefined
Genomic DNA isolation from plants, OmniPrep™
Catalog # 786-397
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Precipitation
    Organic extraction
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Description:
    OmniPrep™ for Plant
  • Description:
    OmniPrep™ for Plant
  • Yield:
    0.5 - 3 μg/mg plant tissue

Specifications

About this item

OmniPrep™ for Plant Kit isolates high quality genomic DNA from plant samples, including fresh and frozen plant tissues. If used according to the protocols this kit purifies DNA from 20gm plant tissue.

  • High Yield, ~100 kb genomic DNA
  • A₂₆₀/A₂₈₀ between 1,8 and 2,0
  • Simple two tube method, 20 to 40 minutes
  • Overcomes the high concentration of plant polysaccharides
  • No toxic chemicals, no phenol, no hazardous waste