Order Entry
Belgium
ContactUsLinkComponent
Genomic DNA isolation from blood, OmniPrep™ for Blood kit
Genomic DNA isolation from blood, OmniPrep™ for Blood kit
Catalog # 786-396
Supplier:  G-Biosciences
CAS Number:  
undefined
Genomic DNA isolation from blood, OmniPrep™ for Blood kit
Catalog # 786-396
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Precipitation
    Organic extraction
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
  • Target molecule:
    DNA
  • Description:
    OmniPrep™ for Blood
  • Description:
    OmniPrep™ for Blood
  • Yield:
    5 - 30 µg/ml blood

Specifications

About this item

OmniPrep™ for Blood Kit isolates high quality genomic DNA from blood samples, including whole blood, buffy coats, packed cells and bone marrow.

  • High Yield, ~100 kb genomic DNA
  • A₂₆₀/A₂₈₀ between 1,8 and 2,0
  • Simple two tube method, 20 to 40 minutes
  • No toxic chemicals, no phenol, no hazardous waste