Pycnometers for bituminous materials
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
About this item
Soda-lime glass or borosilicate glass 3.3.
- Two-part assembly both parts with identification number
- Cap with funnel for ease of filling
- Ring mark on cap stem
4 Options Available Below
Product Details & Documents
Soda-lime glass or borosilicate glass 3.3.
- Two-part assembly both parts with identification number
- Cap with funnel for ease of filling
- Ring mark on cap stem
Product Family Options
Complete sets
Product Information
- Volume (nom.)NSAvailabilityPrice
- NEUB0321-50-1000Pycnometer with ring mark, matt shield and numbering, soda galssPycnometer with ring mark, matt shield and numbering, soda galss1000 ml60Log in for availability1 * 1 ST190,00 €
- NEUB0321-51-1000Pycnometer with ring mark, matt shield and numbering, borosilicate glass 3.3Pycnometer with ring mark, matt shield and numbering, borosilicate glass 3.31000 ml60Log in for availability1 * 1 ST329,00 €
Attachments only
Product Information
- Volume (nom.)NSAvailabilityPrice
- NEUB0321-60-60Top attachment with ring mark, without numbering
Bottles only
Product Information
- Volume (nom.)NSAvailabilityPrice
- NEUB0321-50-0011Bottle with numbering and matt shield, without top, borosilicate glass 3.3Bottle with numbering and matt shield, without top, borosilicate glass 3.31000 ml60/46Log in for availability1 * 1 ST198,00 €