Bouffant caps, PP
Supplier: Hubei Orient International
About this item
Disposable bouffant caps made from non woven PP. Ideal for use in non hazardous environments.
- Single elastic band
- Latex-free
- Soft and comfortable
Weight: 16 to 23 g/m²
: 100 pieces per polybag. 2000 pieces per carton.
5 Options Available Below
Product Details & Documents
Disposable bouffant caps made from non woven PP. Ideal for use in non hazardous environments.
- Single elastic band
- Latex-free
- Soft and comfortable
Weight: 16 to 23 g/m²
: 100 pieces per polybag. 2000 pieces per carton.
Specifications for Product Family
Specifications
- Catalog No:
- 113-9222
- 113-9224
- 113-8133
- 113-9225
- 113-9226
- Garment application:
- Medical|Clinical
- Medical|Clinical
- Medical|Clinical
- Medical|Clinical
- Medical|Clinical
Specifications
Product Family Options
Product Information
- ColourØAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-9222Garment applicationMedical|Clinical - Specifications:
More
Specifications
Cat. No.113-9224Garment applicationMedical|Clinical - Specifications:
More
Specifications
Cat. No.113-8133Garment applicationMedical|Clinical - Specifications:
More
Specifications
Cat. No.113-9225Garment applicationMedical|Clinical - Specifications:
More
Specifications
Cat. No.113-9226Garment applicationMedical|Clinical



