Orders LinkContactUsLinkComponent
Sample cups for analysers
Sample cups for analysers
Supplier:   PerkinElmer
Sample cups for analysers
Supplier:  PerkinElmer
Certificates
Sample cups for analysers
5 Options Available Below
Product Details & Documents
Disposable.
These sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample. For concentrated acids including HF for analyses.
Product Family Options
Product Information
  • Capacity
    Colour
    Availability
    Price
  • PERCB0510397
    Sample cups for auto sampler, PP, no known solvent at room temperature
    Sample cups for auto sampler, PP, no known solvent at room temperature
    1,2 ml
    Translucent
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 243,00 €
    243,00 €
    1 * 2.000 ST
       243,00 €
  • PERCB3001566
    Sample cups for auto sampler, PP, no known solvent at room temperature
    Sample cups for auto sampler, PP, no known solvent at room temperature
    2,5 ml
    Translucent
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 176,00 €
    176,00 €
    1 * 1.000 ST
       176,00 €
  • PERCB3001567
    Sample cups for auto sampler, PP, recommended for blank solutions, reagent solutions and matrix modifiers
    Sample cups for auto sampler, PP, recommended for blank solutions, reagent solutions and matrix modifiers
    7 ml
    Translucent
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 81,00 €
    81,00 €
    1 * 100 ST
       81,00 €
  • PERCB0087600
    Sample cups for auto sampler, PTFE, recommended for use with most concentrated acids including HF and for analyses where the lowest detection levels are required
    Sample cups for auto sampler, PTFE, recommended for use with most concentrated acids including HF and for analyses where the lowest detection levels are required
    2 ml
    Clear
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 831,00 €
    831,00 €
    1 * 30 ST
       831,00 €
  • PERCB0119079
    Sample cups for auto sampler, PS, recommended for use with most aqueous solutions
    Sample cups for auto sampler, PS, recommended for use with most aqueous solutions
    1,5 - 2 ml
    Clear
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 257,00 €
    257,00 €
    1 * 1.000 ST
       257,00 €