Bone curettes, Aesculap®
Supplier: Aesculap
Jump to:
About this item
51 Options Available Below
Product Details & Documents
Product Family Options
Bruns
Product Information
- LengthAvailabilityPrice
- AESCFK611RBone curette, Bruns type (Fig. 000), with 3 mm end
- AESCFK612RBone curette, Bruns type (Fig. 00), with 4 mm end
- AESCFK613RBone curette, Bruns type (Fig. 0), with 5 mm end
- AESCFK614RBone curette, Bruns type (Fig. 1), with 6 mm end
- AESCFK615RBone curette, Bruns type (Fig. 2), with 8 mm end
- AESCFK616RBone curette, Bruns type (Fig. 3), with 10 mm end
- AESCFK617RBone curette, Bruns type (Fig. 4), with 12 mm end
Schede
Product Information
- LengthAvailabilityPrice
- AESCFK621RBone curette, Schede type (Fig. 000), with 2,5 mm end
- AESCFK622RBone curette, Schede type (Fig. 00), with 3,4 mm end
- AESCFK623RBone curette, Schede type (Fig. 0), with 4,2 mm end
- AESCFK624RBone curette, Schede type (Fig. 1), with 5,2 mm end
- AESCFK625RBone curette, Schede type (Fig. 2), with 6,3 mm end
- AESCFK626RBone curette, Schede type (Fig. 3), with 7,4 mm end
- AESCFK627RBone curette, Schede type (Fig. 4), with 8,0 mm end
Volkmann
Product Information
- LengthAvailabilityPrice
- AESCFK631RBone curette, Volkmann type (Fig. 000), with 3,6 mm end
- AESCFK632RBone curette, Volkmann type (Fig. 00), with 4,4 mm end
- AESCFK633RBone curette, Volkmann type (Fig. 0), with 5,2 mm end
- AESCFK634RBone curette, Volkmann type (Fig. 1), with 6,8 mm end
- AESCFK635RBone curette, Volkmann type (Fig. 2), with 8,5 mm end
- AESCFK636RBone curette, Volkmann type (Fig. 3), with 10 mm end
- AESCFK637RBone curette, Volkmann type (Fig. 4), with 11,5 mm end
- AESCFK638RBone curette, Volkmann type (Fig. 5), with 13 mm end
- AESCFK639RBone curette, Volkmann type (Fig. 6), with 14,5 mm end
Williger
Product Information
- LengthAvailabilityPrice
- AESCFK658RBone curette, Willinger type (Fig. 00), with 3,4 mm end
- AESCFK659RBone curette, Willinger type (Fig. 1), with 5,2 mm end
- AESCFK814RBone curette, Willinger type, double ended
Uffenorde
Product Information
- LengthAvailabilityPrice
- AESCFK660RBone curette, type Uffenrode, curved to the left
- AESCFK661RBone curette, type Uffenrode, curved to the right
Daubenspeck
Product Information
- LengthAvailabilityPrice
- AESCFK662RBone curette, type Daubenspeck (Fig. 0000), with 2,8 mm end, angled upwardsBone curette, type Daubenspeck (Fig. 0000), with 2,8 mm end, angled upwards200 mmLog in for availability1 * 1 ST
- AESCFK663RBone curette, type Daubenspeck (Fig. 000), with 3,6 mm end, angled upwardsBone curette, type Daubenspeck (Fig. 000), with 3,6 mm end, angled upwards200 mmLog in for availability1 * 1 ST
- AESCFK664RBone curette, type Daubenspeck (Fig. 00), with 4,4 mm end, angled upwardsBone curette, type Daubenspeck (Fig. 00), with 4,4 mm end, angled upwards200 mmLog in for availability1 * 1 ST
- AESCFK665RBone curette, type Daubenspeck (Fig. 0), with 5,2 mm end, angled upwardsBone curette, type Daubenspeck (Fig. 0), with 5,2 mm end, angled upwards200 mmLog in for availability1 * 1 ST
Lempert
Product Information
- LengthAvailabilityPrice
- AESCFK675RBone curette, type Lempert (Fig. 0000), with 1,8 mm end
- AESCFK676RBone curette, type Lempert (Fig. 000), with 2,0 mm end
- AESCFK677RBone curette, type Lempert (Fig. 00), with 2,4 mm end
- AESCFK678RBone curette, type Lempert (Fig. 0), with 2,6 mm end
- AESCFK679RBone curette, type Lempert (Fig. 1), with 2,8 mm end
Halle
Product Information
- LengthAvailabilityPrice
- AESCFK684RBone curette, type Halle (Fig. 00), with 4,4 mm end, flexible shaftBone curette, type Halle (Fig. 00), with 4,4 mm end, flexible shaft215 mmLog in for availability1 * 1 ST
- AESCFK685RBone curette, type Halle (Fig. 0), with 5,2 mm end, flexible shaftBone curette, type Halle (Fig. 0), with 5,2 mm end, flexible shaft216 mmLog in for availability1 * 1 ST
- AESCFK686RBone curette, type Halle (Fig. 1), with 6,8 mm end, flexible shaftBone curette, type Halle (Fig. 1), with 6,8 mm end, flexible shaft218 mmLog in for availability1 * 1 ST
Simon
Product Information
- LengthAvailabilityPrice
- AESCFK688RBone curette, type Simon (Fig. 000), with 3,6 mm end
- AESCFK689RBone curette, type Simon (Fig. 00), with 4,4 mm end
- AESCFK690RBone curette, type Simon (Fig. 0), with 5,2 mm end
Martini
Product Information
- LengthAvailabilityPrice
- AESCFK800RBone curette, type Martini, double ended (small)
- AESCFK801RBone curette, type Martini, double ended (large)
Jansen
Product Information
- LengthAvailabilityPrice
- AESCFK803RBone curette, type Jansen, double ended
Jacobson
Product Information
- LengthAvailabilityPrice
- AESCFK813RBone curette, type Jacobson, double ended
With PEEK-handle
Product Information
- LengthAvailabilityPrice
- AESCFK666RBone curette with PEEK-handle, Fig. 00, with 4,4 mm end
- AESCFK668RBone curette with PEEK-handle, Fig. 1, with 6,8 mm end
- AESCFK670RBone curette with PEEK-handle, Fig. 2, with 8,5 mm end
- AESCFK672RBone curette with PEEK-handle, Fig. 3, with 10 mm end



