Boiling baths, Precisterm
Supplier: SELECTA
Jump to:
About this item
Double skinned AISI 310 stainless steel tank with an exterior case of AISI 304 stainless steel. The heating elements embedded in INCOLOY heat and corrosion resistant stainless steel.
- Illuminated mains power switch
- Hydraulic thermostat temperature control, synchronised with the 'ON' indicator lamp
- Safety thermostat cut out indicator lamp (DIN 12877.2)
4 Options Available Below
Product Details & Documents
Double skinned AISI 310 stainless steel tank with an exterior case of AISI 304 stainless steel. The heating elements embedded in INCOLOY heat and corrosion resistant stainless steel.
- Illuminated mains power switch
- Hydraulic thermostat temperature control, synchronised with the 'ON' indicator lamp
- Safety thermostat cut out indicator lamp (DIN 12877.2)
Product Family Options
Product Information
- TypeTemp.CapacityPowerBath W×D×HW×D×HWeightAvailabilityPrice
- 6000141Max. 110 °C20 L1500 W480×300×150 mm580×420×200 mm10 kgLog in for availability1 * 1 ST723,93 €
- 6000142 with built in drain tapMax. 110 °C45 L2400 W620×500×150 mm690×620×220 mm18 kgLog in for availability1 * 1 Pce
- 6000386Max. 200 °C5 L1110 W300×140×150 mm370×270×200 mm6 kgLog in for availability1 * 1 ST570,06 €
- 6000388Max. 200 °C20 L2100 W480×300×150 mm580×420×200 mm10 kgLog in for availability1 * 1 ST772,94 €
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- Flat lid with concentric reduction rings, 12 placesFlat lid with concentric reduction rings, 12 placesSELE600160345 L bathLog in for availability1 * 1 ST - 376,80 €
Product Information
- Catalog #ForAvailabilityPrice
- Flat lid with concentric reduction rings, 2 placesFlat lid with concentric reduction rings, 2 placesSELE60032325 L bathLog in for availability1 * 1 ST - 111,71 €
Product Information
- Catalog #ForAvailabilityPrice
- Flat lid with concentric reduction rings, 4 placesFlat lid with concentric reduction rings, 4 placesSELE600323412 L bathLog in for availability1 * 1 Pce - 164,89 €
Product Information
- Catalog #ForAvailabilityPrice
- Flat lid with concentric reduction rings, 6 placesFlat lid with concentric reduction rings, 6 placesSELE600323620 L bathLog in for availability1 * 1 ST - 188,41 €
Product Information
- Catalog #ForAvailabilityPrice
- Gabled stainless steel lid with thermometer positioning holeGabled stainless steel lid with thermometer positioning holeSELE600071220 L bathLog in for availability1 * 1 Pce - 206,54 €
Product Information
- Catalog #ForAvailabilityPrice
- Gabled stainless steel lid with thermometer positioning holeGabled stainless steel lid with thermometer positioning holeSELE600071445 L bathLog in for availability1 * 1 ST - 345,29 €
Product Information
- Catalog #ForAvailabilityPrice
- Gabled stainless steel lid with thermometer positioning holeGabled stainless steel lid with thermometer positioning holeSELE600071112 L bathLog in for availability1 * 1 ST - 166,93 €
Product Information
- Catalog #ForAvailabilityPrice
- Gabled stainless steel lid with thermometer positioning holeGabled stainless steel lid with thermometer positioning holeSELE60007105 L bathLog in for availability1 * 1 Pce - 142,20 €