Orders LinkContactUsLinkComponent
Gel analysis software, Dymension
Gel analysis software, Dymension
Gel analysis software, Dymension
Certificates
Gel analysis software, Dymension
3 Options Available Below
Product Details & Documents
Dymension software can be used to analyse a typical 2D gel image rapidly. It features algorithms for background subtraction, noise filtering, precise alignment, spot detection, rapid matching and reduced image editing time. Using its powerful spot detection algorithm, Dymension instantly locates and analyses protein spots. With Dymension, the entire analysis process from background correction to spot matching results and reporting takes minutes. Dymension is available in three versions, and upgrading between the packages is simple.
Dymension 1 is ideal for users looking for software to analyse spots between samples containing one 2D gel only. It offers a range of automatic image correction features and methods of resolving and analysing spots on single gels. The software can be easily upgraded to Dymension 2 or 3 if the proteomics needs of the laboratory change.
Dymension 2 has all the features of Dymension 1 but has the added ability to detect and compare protein spot maps in samples containing replicate gels. The software offers many background correction and noise filtering features including a SYPRO® Ruby filter, which can detect and automatically remove spots caused by SYPRO® Ruby crystals adhering to the gel, thus saving users valuable time editing their image. Dymension 2 is suitable for users that need to determine the amount of protein present before and after drug or protein induction treatments. The software automatically detects and assigns statistical confidence to each and every difference in spot normalised volume, thus accurately highlighting proteins of specific interest. The results of the analysis are displayed as a dynamically linked table, 3D spot profile, bar chart, correlation and scatter plot, which make it easy to compare many expression profiles simultaneously and therefore accurately pick proteins suitable for further analysis.
Dymension 3 has all the functionality of Dymension 2 so can be used to analyse samples containing replicate gels. Dymension 3 has the additional capability to analyse multi-stained fluorescent gel images stained with up to three contrasting labels, eg, Cy™2, Cy™3 and Cy™5. This feature allows users to analyse multiple sets of any type of 2D protein gel, regardless of how it was stained, using only one software package.
Product Family Options
Product Information
  • For
    Availability
    Price
  • 730-1161
    Dymension 1
    Dymension 1
    Analysis of single 2D gels
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 080,00 €
    2 080,00 €
    1 * 1 ST
       2 080,00 €
  • 730-1162
    Dymension 2
    Dymension 2
    Rapid analysis of multiple 2D gel sets
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 740,00 €
    3 740,00 €
    1 * 1 ST
       3 740,00 €
  • 730-1163
    Dymension 3
    Dymension 3
    Quick and easy analysis of all 2D gel applications
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 200,00 €
    5 200,00 €
    1 * 1 ST
       5 200,00 €