Order Entry
Belgium
ContactUsLinkComponent
Spray caps for dropping bottles
Spray caps for dropping bottles
  MUEL14.197.01
undefined
Spray caps for dropping bottles
  MUEL14.197.01

 

  • For:
    GL 18 thread
  • Description:
    Spray cap, size 40 mm, disperse 0,14 ml of liquid per drive
  • Pk:
    20
  • For bottles:
    10/100 ml

 

 

The spray caps with DIN 18 ring and protective cap are suitable for use with 10 to 100 ml dropping bottles.