Orders LinkContactUsLinkComponent
Protein blocking solution, BLOK™ BLOTTO
Protein blocking solution, BLOK™ BLOTTO
BLOK™ BLOTTO 5% non fat milk solution, 2
Catalog # 786-192
Supplier:   G-Biosciences
Protein blocking solution, BLOK™ BLOTTO
Catalog # 786-192
Supplier:  G-Biosciences
CAS Number:  
Protein blocking solution, BLOK™ BLOTTO
Protein blocking solution, BLOK™ BLOTTO
BLOK™ BLOTTO 5% non fat milk solution, 2
CAS Number:  
Price
184,00 €
Log in for availability
1 * 2 ST
Product Details & Documents
BLOK™ BLOTTO is a 5% novel modified milk protein solution used for Westerns, ELISA and dot blot blocking.
  • Ready to use
  • Blocking in 10 to 15 mins
Documents
Specifications
Specifications
  • Pk:
    2
  • Description:
    BLOK™ BLOTTO 5% non fat milk solution