Orders LinkContactUsLinkComponent
PCR microtubes
PCR microtubes
PCR tubes, individual, with attached cap, Clear, 0,5 ml
Catalog # 731-0407
   Low Manufacturing Impact
Supplier:   EPPENDORF
PCR microtubes
Catalog # 731-0407
Supplier:  EPPENDORF
   Low Manufacturing Impact
PCR microtubes
PCR microtubes
PCR tubes, individual, with attached cap, Clear, 0,5 ml
Price
56,70 €
Log in for availability
1 * 500 ST
Product Details & Documents
PP.
  • Etched lid for labelling
  • Tight sealing
  • Easy to open
  • Certified free from human DNA, DNase, RNase and PCR inhibitors
These thin-walled, 0,5 ml PCR tubes are designed for use with all thermal cyclers with 0,5 ml block format.
Colour: Translucent
Documents
Specifications
  • Capacity:
    0,5 ml
  • Colour:
    Clear
  • Description:
    PCR tubes, individual, with attached cap
  • Cat. No.:
    731-0407
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact