Order Entry
Belgium
Orders LinkContactUsLinkComponent
Conductivity sensor, inductive, 3700 SC

Conductivity sensor, inductive, 3700 SC

Conductivity sensor, conductive, sanitary style, 3700 SC, PFA, 50,8×127 mm
Catalog # HACHD3708E2T.99
Supplier:   Hach
Conductivity sensor, inductive, 3700 SC
Catalog # HACHD3708E2T.99
Supplier:  Hach
Conductivity sensor, inductive, 3700 SC
Conductivity sensor, inductive, 3700 SC

Conductivity sensor, conductive, sanitary style, 3700 SC, PFA, 50,8×127 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

The innovative technology of Hach’s inductive conductivity sensors eliminates polarization and electrode coating problems in harsh environments. The sensor comes complete with standard sensor cable length of 6 m, digital gateway, and a 1 m digital extension cable.

  • Sanitary (CIP) flange style: 2-inch flange, special cap, and EPDM compound gasket
  • Conforms to provisions of 3-A Sanitary Standards
Specifications
  • Ø×L:
    50,8×127 mm
  • Type:
    3700 SC
  • Description:
    Conductivity sensor, conductive, sanitary style
  • Material:
    PFA
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.