Rack for COD reagent vials
10 (1×10), COD, Stainless steel
N° de catalogue 705-0078
Fournisseur: Hach
Jump to:
Product Details & Documents
Stainless steel.
Rack for holding hot COD vials or test tubes.
Specifications
- For tubes:COD
- No. of Holes:10 (1×10)
- Material:Stainless steel