Non-animal protein blocking Agent, NAP-BLOCKER™
Fournisseur: G-Biosciences
Certificats
À propos de cet article
NAP-BLOCKER™ is an animal-free protein blocking agent recommended for improved assay sensitivity, minimal non specific binding and a high signal-to-background ratio. It is suitable for Western blots, dot blots and assay development.
- Free from animal source blocking agents
- Eliminates cross reactivity
- Eliminates non specific binding
Frequently Bought Together