Orders LinkContactUsLinkComponent
Extraction buffers, FOCUS™
Extraction buffers, FOCUS™
Extraction buffers, FOCUS™
Extraction buffers, FOCUS™
FOCUS™ Extraction Buffers are experimentally optimised concentrations of critical agents, buffering and stabilising agents, including urea, thiourea, Nonidet® P-40, CHAPS, and sulphobetaines (SB). They are designed to produce optimal protein extraction and improved spot resolution for 2D gel analysis.
Product Family Options
  • Description
    Pk
  • FOCUS™ Extraction Buffer I, includes urea and Nonidet® P-40
    50 mL
    1 * 50 mL
    1 * 50 mL - 143,00 €
    143,00 €
    1 * 50 mL
       143,00 €
    Specifications
    Cat. No.
    786-220
  • FOCUS™ Extraction Buffer II, includes urea and CHAPS
    50 mL
    1 * 50 mL
    1 * 50 mL - 213,00 €
    213,00 €
    1 * 50 mL
       213,00 €
    Specifications
    Cat. No.
    786-221
  • FOCUS™ Extraction Buffer III, includes urea, thiourea, CHAPS and amidosulfobetaine (ASB)-16
    50 mL
    1 * 50 mL
    1 * 50 mL - 213,00 €
    213,00 €
    1 * 50 mL
       213,00 €
    Specifications
    Cat. No.
    786-222
  • FOCUS Extraction Buffer IV, includes urea, thiourea, CHAPS and sulphobetaine (SB) 3-10
    50 mL
    1 * 50 mL
    1 * 50 mL - 213,00 €
    213,00 €
    1 * 50 mL
       213,00 €
    Specifications
    Cat. No.
    786-223
  • FOCUS™ Extraction Buffer V, includes urea, thiourea and CHAPS
    1 KIT
    1 * 1 KIT
    1 * 1 KIT - 213,00 €
    213,00 €
    1 * 1 KIT
       213,00 €
    Specifications
    Cat. No.
    786-219
  • FOCUS™ Extraction Buffer VI, includes urea, thiourea, CHAPS and non detergent sulphobetaine 201
    50 mL
    1 * 50 mL
    1 * 50 mL - 213,00 €
    213,00 €
    1 * 50 mL
       213,00 €
    Specifications
    Cat. No.
    786-233
  • FOCUS™ Extraction Buffers I-VI trial kit (includes each 10 ml)
    1 KIT
    1 * 1 KIT
    1 * 1 KIT - 372,00 €
    372,00 €
    1 * 1 KIT
       372,00 €
    Specifications
    Cat. No.
    786-234
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.