Order Entry
Belgium
ContactUsLinkComponent
 
Genomic DNA isolation from tissue, Axygen® AxyPrep™ Mag
 :  Corning
 
undefined
Genomic DNA isolation from tissue, Axygen® AxyPrep™ Mag
 :  Corning
 

 

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    AxyPrep Mag Tissue gDNA, small

 

 

Frequently Bought Together