Deep well plates, 24-well, Axygen®
Fournisseur: Corning
Certificats
À propos de cet article
Deep well and assay plates are used for sample collection, in vitro growth chambers and longterm storage. Made of PP, they offer excellent chemical resistance and will withstand temperatures down to –80 °C. Ultra-flat surface ensures proper sealing with heat sealing film. Clear V-shaped well bottom.
Spécifications
- Catalog No:
- AXYGPDW10ML24C2BCS
- AXYGP-DW10ML24CS
- AXYGP-DW10ML24C
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
Spécifications
Frequently Bought Together