ContactUsLinkComponent
Magnetic stirring bars positioner/retrievers, Spinbar®, SP Bel-Art
Catalog # BELAH377800000
undefined
Magnetic stirring bars positioner/retrievers, Spinbar®, SP Bel-Art
Catalog # BELAH377800000
Spécifications
  • Ø:
    15,9 mm
  • Length:
    305 mm
  • Pk:
    1
Spécifications
À propos de cet article
PE, to retrieve stirbars without coming into contact with the liquid in the containers.
  • No direct contact with stirring magnet
  • Splash-free operation