Buffers for Biacore Systems
Supplier: Cytiva
Certificates
About this item
Buffers developed and verified to work in Biacore™ systems.
Immobilisation buffers
A range of buffers for the most common ligand types and immobilisation conditions.
Regeneration buffers
Regeneration is the step where bound analyte is removed from the sensor chip after analysis, without affecting the activity of the immobilised ligand. In many systems, conditions that remove analyte tend to reduce ligand activity, and finding the optimal conditions is an essential part of assay development.
Related Information
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together