Spin Adapters for Gravity Columns
About this item
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
- Convenient adapters, enable spinning of all gravity columns
Adapters are polypropylene rings designed for use with the gravity columns PD MiniTrap G-25, PD MidiTrap G-25, and PD-10 Desalting Columns when the spin protocol is used. MiniSpin Adapter fits PD MiniTrap G-25 columns, MidiSpin Adapter fits PD MidiTrap G-25 columns, and PD-10 Spin Adapter fits PD-10 Desalting Columns.
3 Options Available Below
Product Details & Documents
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
- Convenient adapters, enable spinning of all gravity columns
Adapters are polypropylene rings designed for use with the gravity columns PD MiniTrap G-25, PD MidiTrap G-25, and PD-10 Desalting Columns when the spin protocol is used. MiniSpin Adapter fits PD MiniTrap G-25 columns, MidiSpin Adapter fits PD MidiTrap G-25 columns, and PD-10 Spin Adapter fits PD-10 Desalting Columns.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Product Family Options
Product Information
- AvailabilityPrice
- 28-9232-43MiniSpin™ adapter, for PD MiniTrap™ G-25 columns
- 28-9232-44MidiSpin™ adapter, for PD MidiTrap™ G-25 columns
- 28-9232-45PD-10 Spin adapter, for PD-10 desalting columns
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.