Order Entry
Belgium
ContactUsLinkComponent
DNA (Deoxyribonucleic acid) (from calf thymus), sonicated
DNA (Deoxyribonucleic acid) (from calf thymus), sonicated
Catalog # 27-4563-01
Supplier:  Cytiva
CAS Number:  
undefined
DNA (Deoxyribonucleic acid) (from calf thymus), sonicated
Catalog # 27-4563-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0345
  • Pk Info:
    100 OD

Specifications

About this item

 :
Storage Temperature:Freezer
CAS Number:9007-49-2
 :73049-39-5
MDL Number:MFCD00130922