Connection system GL 45
Over dit artikel
PP screwcaps for Pyrex® GL 45 media-lab bottles facilitate the transfer of liquids within a closed and sterile system.
- Temperature resistant to 140 °C
- Membrane screw cap is ideal for aerobic microbiology and cell culture, with gas permeable
- Dishwasher safe
Ideal for chemistry and biotechnology laboratories. Components are fully interchangeable and compatible with both two and three port screwcaps. Four different tubing diameters can be accommodated (Ext.Ø; 1,6, 3,0, 3,2 and 6,0 mm). The additional pressure compensation set that includes a membrane filter with 0,2 μm sized pores permits sterile pressure equalisation. Unused ports can be sealed with a blanking cap. Components are fully interchangeable and compatible with both two and three port screwcaps.
14 Opties Hieronder Beschikbaar
Product Details & Documents
PP screwcaps for Pyrex® GL 45 media-lab bottles facilitate the transfer of liquids within a closed and sterile system.
- Temperature resistant to 140 °C
- Membrane screw cap is ideal for aerobic microbiology and cell culture, with gas permeable
- Dishwasher safe
Ideal for chemistry and biotechnology laboratories. Components are fully interchangeable and compatible with both two and three port screwcaps. Four different tubing diameters can be accommodated (Ext.Ø; 1,6, 3,0, 3,2 and 6,0 mm). The additional pressure compensation set that includes a membrane filter with 0,2 μm sized pores permits sterile pressure equalisation. Unused ports can be sealed with a blanking cap. Components are fully interchangeable and compatible with both two and three port screwcaps.
Product Family Options
Connection system
Productinformatie
Twin tubing connectors
Productinformatie
Membrane screw caps
Productinformatie
- DescriptionBeschikbaarheidPrice
- Membrane screwcap, PP, blue, GL 45, with 0,2 μm PTFE membraneLog in voor beschikbaarheid1 * 5 ST42,90 €
Bottle pourer
Productinformatie
- DescriptionBeschikbaarheidPrice
- Bottle pourer, PTFE, GL 45, with venting tube and attached PP blue capLog in voor beschikbaarheid1 * 1 ST151,00 €