Order Entry
Belgium
ContactUsLinkComponent
 
Bi-metallic dial thermometer with Min/Max memory
  MOIN4100T
 :  MOINEAU
undefined
Bi-metallic dial thermometer with Min/Max memory
  MOIN4100T
 :  MOINEAU

 

  • Ø:
    114 mm
  • Division:
    1 °C
  • Pk:
    1
  • Range:
    −40…+50 °C
  • Weight:
    80 g

 

 

Maximum/minimum dial thermometer, bi-metallic.

  • For wall mounting
  • Made from white ABS

Two markers show the maximum and minimum temperature until the next reset.