ContactUsLinkComponent
Delivery tip for titration, anti-diffusion, D4353
Delivery tip for titration, anti-diffusion, D4353
Catalog # HACH956-309
Supplier:  Hach
undefined
Delivery tip for titration, anti-diffusion, D4353
Catalog # HACH956-309
Supplier:  Hach
Specifications
  • Description:
    Anti-diffusion delivery tip, D4353, 100 mm length
  • Pk:
    1
Specifications
About this item
PTFE titrant delivery tip for use mounted on delivery tubing.