ContactUsLinkComponent
Thermostatic baths for viscometers, CT 72
Thermostatic baths for viscometers, CT 72
Supplier:  SI Analytics
Certificates
undefined
Thermostatic baths for viscometers, CT 72
Supplier:  SI Analytics
Specifications
  • Catalog No:
  • 645-0081
  • SCOI285418547
  • SCOI285418615
  • SCOI285418526
  • 645-0080
  • Model:
  • CT 72/4
  • CT 72/2
  • CT 72/2-TT
  • CT 72/P
  • CT 72/E
  • Capacity:
  • 27 L
  • 15 L
  • 15 L
  • 15 L
  • 15 L
  • Temperature range:
  • 5...150 °C
  • 5...150 °C
  • –40...+150 °C
  • 10...60 °C
  • 10...60 °C
  • Viscometer positions:
  • 4
  • 2
  • 2
  • 2
  • 2
  • Bath material:
  • Stainless steel and glass
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • Stainless steel
  • W×D×H:
  • 605×250×370 mm
  • 355×370×250 mm
  • 355×370×250 mm
  • 355×370×250 mm
  • 355×370×250 mm
  • Weight:
  • 28 kg
  • 13,5 kg
  • 14 kg
  • 5 kg
  • 5 kg