Steady-Luc™ Firefly HTS assay kit
: Biotium
Firefly luciferase is widely used as a transcriptional reporter gene for research and drug screening. The Steady-Luc™ HTS kit can be used with luminescence plate readers without sample injection systems or for high-throughput screening.
Frequently Bought Together