Orders LinkContactUsLinkComponent
Slide box
Slide box
Box for microscope preparations, Blue, Plastic, 25
Catalog # LIEDPK25
Supplier:   LIEDER JOHANNES
Slide box
Catalog # LIEDPK25
Supplier:  LIEDER JOHANNES
Slide box
Slide box
Box for microscope preparations, Blue, Plastic, 25
Price
5,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
This slide box is designed to facilitate the transportation of 25 microscope slides.
  • Transparent cover for easy viewing
  • Robust design and stackable construction
  • Serrated retainer strips to maintain slides properly
Specifications
  • No. of slides:
    25
  • Colour:
    Blue
  • Description:
    Box for microscope preparations
  • Material:
    Plastic