Orders LinkContactUsLinkComponent
Detectable plaster rolls
Detectable plaster rolls
Supplier:   SOEHNGEN
Detectable plaster rolls
Supplier:  SOEHNGEN
Certificates
Detectable plaster rolls
3 Options Available Below
Product Details & Documents
These detectable plasters are used to cover small skin lesions. The aluminium strip under the wound pad make the plasters detectable by all conventional metal detectors. Ideal for certain areas of production, in particular the food industry.
  • Skin-friendly and reliable acrylic adhesive
  • Water resistant, hypoallergenic and breathable
  • Blue colour ensures easy visual recognition
Product Family Options
Product Information
  • L×W
    Availability
    Price
  • SOEH1009090
    Detectable plaster roll, blue, 1 m
    Detectable plaster roll, blue, 1 m
    1000×60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,90 €
    13,90 €
    1 * 1 ST
       13,90 €
  • SOEH1009091
    Detectable plaster roll, blue, 1 m
    Detectable plaster roll, blue, 1 m
    1000×80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • SOEH1009998
    Detectable plaster roll, 5 m
    Detectable plaster roll, 5 m
    5000×60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST