Readybag® Granulated Media for Salmonella and Listeria Control
Supplier: Merck
Certificates
About this item
Readybag® gamma-irradiated and pre-weighed granulated media pouches for fast, flexible and easy pathogen testing. Readybag® media pouches speed up and simplify food pathogen testing routines: All media preparation steps are eliminated from workflow. Just add sterile water before incubation. Readybag® series includes Salmonella (Buffered Peptone Water) and Listeria (Half Fraser Broth).
- No weighing
- No autoclaving
- No supplement handling
- No need for extra lab space or equipment
Frequently Bought Together