Orders LinkContactUsLinkComponent
Dipping Pipettes
Dipping Pipettes
Certificates
Dipping Pipettes
5 Options Available Below
Product Details & Documents
Dipping pipettes for sampling predefined volumes. The pipettes are immersed into the samples and filled via the open top. The pipette is emptied by tilting sideways. Calibrated to deliver EX.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • NEUB0276-50-005
    Dipping pipette, tolerance ±0,02 ml
    Dipping pipette, tolerance ±0,02 ml
    5 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,60 €
    65,60 €
    1 * 1 ST
       65,60 €
  • NEUB0276-50-010
    Dipping pipette, tolerance ±0,03 ml
    Dipping pipette, tolerance ±0,03 ml
    10 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 85,20 €
    85,20 €
    1 * 1 ST
       85,20 €
  • NEUB0276-50-025
    Dipping pipette, tolerance ±0,06 ml
    Dipping pipette, tolerance ±0,06 ml
    25 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 96,40 €
    96,40 €
    1 * 1 ST
       96,40 €
  • NEUB0276-50-050
    Dipping pipette, tolerance ±0,1 ml
    Dipping pipette, tolerance ±0,1 ml
    50 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 97,20 €
    97,20 €
    1 * 1 ST
       97,20 €
  • NEUB0276-50-100
    Dipping pipette, tolerance ±0,2 ml
    Dipping pipette, tolerance ±0,2 ml
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 132,00 €
    132,00 €
    1 * 1 ST
       132,00 €