Orders LinkContactUsLinkComponent
Sample concentrators, Reacti Vap™
Sample concentrators, Reacti Vap™
Sample concentrators, Reacti Vap™
Certificates
Sample concentrators, Reacti Vap™
2 Options Available Below
Product Details & Documents
Reacti-Vap Evaporators (9 port)
These evaporators are precision machined gassing manifolds. They provide simple, efficient evaporation by allowing the simultaneous or separate delivery of non-reactive pressurised gas to samples. They easily connect to single block Reacti-Therm™ modules.
Reacti-Vap III Evaporators (27 port)
These evaporators triple the number of samples to evaporate. Nine needles attach to each of the three individually regulated chambers. They easily connect to Reacti- Therm™ III modules.
Product Family Options
Product Information
  • Availability
    Price
  • HYPETS-18825
    Reacti-Vap sample concentrator (9 port)
    Reacti-Vap sample concentrator (9 port)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 240,00 €
    1 240,00 €
    1 * 1 ST
       1 240,00 €
  • HYPETS-18826
    Reacti-Vap III sample concentrator (27 port)
    Reacti-Vap III sample concentrator (27 port)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 320,00 €
    2 320,00 €
    1 * 1 ST
       2 320,00 €
Reacti-therm modules
Product Information
  • Catalog #
    Temperature Range
    Description
    No. of blocks
    Availability
    Price
  • Ambient +10...200 °C
    Reacti-Therm™ module with heating function, for 27 port sample concentrator
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 400,00 €
    3 400,00 €
    1 * 1 ST
       3 400,00 €
Product Information
  • Catalog #
    Temperature Range
    Description
    No. of blocks
    Availability
    Price
  • Ambient +10...200 °C
    Reacti-Therm™ module with heating and stirring function, for 27 port sample concentrator
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 490,00 €
    4 490,00 €
    1 * 1 ST
       4 490,00 €
Product Information
  • Catalog #
    Temperature Range
    Description
    No. of blocks
    Availability
    Price
  • Ambient +10...200 °C
    Reacti-Therm™ module with heating function, for 9 port sample concentrator
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 630,00 €
    2 630,00 €
    1 * 1 ST
       2 630,00 €
Product Information
  • Catalog #
    Temperature Range
    Description
    No. of blocks
    Availability
    Price
  • Ambient +10...200 °C
    Reacti-Therm™ module with heating and stirring function, for 9 port sample concentrator
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 430,00 €
    3 430,00 €
    1 * 1 ST
       3 430,00 €
Needles
Product Information
  • Catalog #
    Gauge
    Length
    Description
    Availability
    Price
  • 19
    152 mm
    Stainless steel PTFE coated needles, blunt end, length: 152 mm
    Log in for availability
    Log in for availability
    1 * 9 ST
    1 * 9 ST - 287,00 €
    287,00 €
    1 * 9 ST
       287,00 €
Product Information
  • Catalog #
    Gauge
    Length
    Description
    Availability
    Price
  • 19
    102 mm
    Stainless steel PTFE coated needles, blunt end, length: 102 mm
    Log in for availability
    Log in for availability
    1 * 9 ST
    1 * 9 ST - 248,00 €
    248,00 €
    1 * 9 ST
       248,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Metal rod
    Metal rod
    HYPETS-18831
    Log in for availability
    1 * 1 ST - 70,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Replacement height adjustment knobs
    Replacement height adjustment knobs
    HYPETS-18829
    Log in for availability
    1 * 1 ST - 37,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Luer-lok fitting
    Luer-lok fitting
    HYPETS-18827
    Log in for availability
    1 * 1 ST - 54,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Replacement tube kit
    Replacement tube kit
    HYPETS-18782
    Log in for availability
    1 * 9 ST - 222,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.