Order Entry
Belgium
ContactUsLinkComponent
 
Septa for 22 mm screw top vials
  HEWL8010-0564
 :  Agilent
undefined
Septa for 22 mm screw top vials
  HEWL8010-0564
 :  Agilent

 

  • For:
    10 ml vials
  • Type:
    PTFE/silicone
  • Description:
    Septa
  • Pk:
    100