Cartridge filters, ProcessGard® CR
Fournisseur: ENTEGRIS
Certificats
À propos de cet article
These cartridge filters are widely used in chemical, electronic and semiconductor industries for liquid and gas prefiltration applications. They retain contaminants by means of a multi-stage, graded density design.
- Depth filter design
- Superior retention of colloids and particles ensuring low particle counts in bulk DI water and chemical production
- Excellent chemical and heat resistance
- Manufactured, tested and packaged in an ISO 9001 Quality Systems Standard facility
Membrane: PP; Supports, cage, core, sleeves and endcaps: PP
Maximum differential pressure: 4,83 bar at 25 °C
Maximum operating temperature: 80 °C
Frequently Bought Together