ContactUsLinkComponent
VWR® Q Path® Safemount, Mounting Agents for Microscopy
VWR® Q Path® Safemount, Mounting Agents for Microscopy
Catalog # 00647520.
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Q Path® Safemount, Mounting Agents for Microscopy
Catalog # 00647520.
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Safemount mounting media Q Path®
Specifications
About this item
Safemount Q Path® is a mounting medium, supplied in an PET bottle, which is compatible with Safesolv. It glues the coverglass to the slide and fixes the probe in position after staining.
  • Amber bottle
  • Easily spreadable
  • Easy to sponge away