Spare needles for autosampler syringes
Fournisseur: Agilent
Certificats
À propos de cet article
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Fournisseur: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Fournisseur: VWR Chemicals
VWR®, Nitrile Gloves
Fournisseur: VWR Collection
pH indicator strips, MQuant®, Supelco®
Fournisseur: Merck
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Fournisseur: Merck
Centrifuge tubes
Fournisseur: Brand
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Fournisseur: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Fournisseur: VWR Chemicals
Serological pipettes, total delivery
Fournisseur: Brand
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Fournisseur: Merck
Accessories for needle assemblies
Fournisseur: Agilent
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Fournisseur: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Fournisseur: VWR Chemicals
Valve supplies for HPLC system
Fournisseur: Agilent
Capillaries for LC systems
Fournisseur: Agilent
CAPILLARY ST 0.3X80 SL-SL 1 * 1 items
Fournisseur: Agilent
Transfection reagent for rAAV production in suspension, FectoVIR®-AAV
Fournisseur: Polyplus-transfection