1000 and 1700 series Gastight® syringes for PSD/2 syringe pump
Supplier: HAMILTON BONADUZ
Certificates
About this item
Gastight® syringes for the PSD/2 syringe pump are available with a standard PTFE-tipped plunger and a long life ultra-high molecular weight polyethylene (UHMWPE) plunger tip. The stroke length of a PSD/2 syringe is 60 mm.
Syringe terminations:
TLL, PTFE® Luer Lock
This termination has a PTFE, male Luer taper with nickel plated brass locking hub for use with Kel-F needles, metal hub needles and universal connectors. Also, the TLL is used with Hamilton Diluters/Dispensers, OEM applications and manual operations. Autoclavable when disassembled, except on 25 ml and greater syringes. Repeated autoclaving will shorten syringe life.
Frequently Bought Together