pH probes
Supplier: TESTO
Jump to:
About this item
5 Options Available Below
Product Details & Documents
Product Family Options
Product Information
- pHTemp.ElectrolyteØ×LForAvailabilityPrice
- 663-3215pH probe, glass, for testo 206 pH3, with temperature sensorpH probe, glass, for testo 206 pH3, with temperature sensor0 - 140...80 °CGel12×120 mmGeneral purposesLog in for availability1 * 1 ST265,00 €
- TEST0650.2061Spare pH probe for testo 206 pH1, with NTC, incl. gel storage capSpare pH probe for testo 206 pH1, with NTC, incl. gel storage cap0 - 140...60 °CGel15×3,5 mmLiquidsLog in for availability1 * 1 ST88,00 €
- TEST0650.2062Spare pH probe for testo 206 pH2, with NTC, incl. gel storage capSpare pH probe for testo 206 pH2, with NTC, incl. gel storage cap0 - 140...60 °CGel11×35 mmSemi-solid substancesLog in for availability1 * 1 ST177,00 €
- TEST0650.2064Universal pH probe, plastic, for testo 206 pH3, with temperature sensorUniversal pH probe, plastic, for testo 206 pH3, with temperature sensor0 - 140...80 °CGel12×120 mmGeneral purposesLog in for availability1 * 1 ST224,00 €
- TEST0650.2063Universal pH probe, plastic, for testo 206 pH3, without temperature sensorUniversal pH probe, plastic, for testo 206 pH3, without temperature sensor0 - 14- °CGel12×120 mmGeneral purposesLog in for availability1 * 1 ST116,00 €