Orders LinkContactUsLinkComponent
Spray bottles
Spray bottles
500 ml
Catalog # 215-4232
Supplier:   LAMAPLAST
Spray bottles
Catalog # 215-4232
Supplier:  LAMAPLAST
Spray bottles
Spray bottles
500 ml
Price
10,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
LDPE, translucent.
  • Good chemical resistance, good temperature resistance
  • Spray nozzle adjustment can be varied by rotation
  • Pressureless spray bottle
Spray, choice of fine mist or focused jet spray functions to disperse fluids.
Documents
Specifications
  • Capacity:
    500 ml
  • Seq:
    01