Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Chromatography Vial Closures
Headspace screw cap with 18 mm septa
Headspace screw cap with 18 mm septa
Headspace screw cap with 18 mm septa
Catalog # HEWL5188-2759
Supplier:
Agilent
Expand image
Headspace screw cap with 18 mm septa
Catalog # HEWL5188-2759
Supplier:
Agilent
Specifications
For:
Screw top vials
Septum:
White PTFE/blue silicone
Type:
Silver, magnetic
Description:
Screw top cap with septa, CombiPAL
Pk:
100
Specifications
About this item
Product Details
Product Details