ContactUsLinkComponent
Adhesion slides, X-tra®
Adhesion slides, X-tra®
Catalog # LEIH3800200AE
Supplier:  LEICA HISTOLOGY
undefined
Adhesion slides, X-tra®
Catalog # LEIH3800200AE
Supplier:  LEICA HISTOLOGY
Specifications
  • Colour:
    White
  • Pk:
    72
Specifications
About this item
The X-tra® slides are designed to bond tissue sections and cytology preparations.
  • High quality material for use with slide printers
  • Corners clipped at 45° to reduce glass breakage and fragmentation
  • Permanent positive charged surface: No need of additional adhesives