Orders LinkContactUsLinkComponent
Cooling elements and frames
Supplier:   DELTA T
Cooling elements and frames
Supplier:  DELTA T
Certificates
Cooling elements and frames
11 Options Available Below
Product Details & Documents
Molecular medical elements for different temperature handling of samples ranges from −32 °C to +38 °C.
  • Made of PE and natural in colour
  • Sealing made of laminated aluminium film with a protected cap
  • A liquid crystal thermometer in the elements shows its current temperature
Used for transportation and storage of samples like blood products, homograft’s, cell cultures, stem cells, antibodies, proteins and other temperature sensitive medications.
Inside dimension: 210×110×30 mm
Product Family Options
Tempshell elements
Product Information
  • Colour
    Temperature Range
    Dimensions
    Availability
    Price
  • DLTTA100005DE
    Cooling elements, −30 °C, Tempshell
    Cooling elements, −30 °C, Tempshell
    Orange
    −32...+30 °C
    275×155×60 mm
    210×110×30 mm
    Log in for availability
    Log in for availability
    1 * 1 PAIRE
    1 * 1 PAIRE - 150,00 €
    150,00 €
    1 * 1 PAIRE
       150,00 €
  • DLTTA100011
    Cooling elements, −21 °C, Tempshell
    Cooling elements, −21 °C, Tempshell
    Translucent white
    −23...+19 °C
    275×155×60 mm
    210×110×30 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 141,00 €
    141,00 €
    1 * 2 ST
       141,00 €
  • DLTTA100017DE
    Cooling elements, −18 °C, Tempshell
    Cooling elements, −18 °C, Tempshell
    Translucent white
    −22...−18 °C
    275×155×60 mm
    210×110×30 mm
    Log in for availability
    Log in for availability
    1 * 1 PAIRE
    1 * 1 PAIRE - 108,00 €
    108,00 €
    1 * 1 PAIRE
       108,00 €
  • DLTTA100029DE
    Cooling elements, 4 °C, Tempshell
    Cooling elements, 4 °C, Tempshell
    Blue
    2...8 °C
    275×155×60 mm
    210×110×30 mm
    Log in for availability
    Log in for availability
    1 * 1 PAIRE
    1 * 1 PAIRE - 150,00 €
    150,00 €
    1 * 1 PAIRE
       150,00 €
  • DLTTA100035
    Cooling elements, 22 °C, Tempshell
    Cooling elements, 22 °C, Tempshell
    Green
    20...24 °C
    275×155×60 mm
    210×110×30 mm
    Log in for availability
    Log in for availability
    1 * 1 PAIRE
    1 * 1 PAIRE - 158,00 €
    158,00 €
    1 * 1 PAIRE
       158,00 €
  • DLTTA100041
    Cooling elements, 37 °C, Tempshell
    Cooling elements, 37 °C, Tempshell
    Red
    10...40 °C
    275×155×60 mm
    210×110×30 mm
    Log in for availability
    Log in for availability
    1 * 1 PAIRE
    1 * 1 PAIRE - 108,00 €
    108,00 €
    1 * 1 PAIRE
       108,00 €
Tempshell frames
Product Information
  • Colour
    Temperature Range
    Dimensions
    Availability
    Price
  • DLTTA100006DE
    Cooling frames, −30 °C, Tempshell
    Cooling frames, −30 °C, Tempshell
    Orange
    −32...+30 °C
    Log in for availability
    Log in for availability
    1 * 1 PAIRE
    1 * 1 PAIRE - 150,00 €
    150,00 €
    1 * 1 PAIRE
       150,00 €
  • DLTTA100012
    Cooling frames, −21 °C, Tempshell
    Cooling frames, −21 °C, Tempshell
    Translucent white
    −23...+19 °C
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 150,00 €
    150,00 €
    1 * 2 ST
       150,00 €
  • DLTTA100018DE
    Cooling frames, −18 °C, Tempshell
    Cooling frames, −18 °C, Tempshell
    Translucent white
    −22...−18 °C
    Log in for availability
    Log in for availability
    1 * 1 PAIRE
    1 * 1 PAIRE - 108,00 €
    108,00 €
    1 * 1 PAIRE
       108,00 €
  • DLTTA100030DE
    Cooling frames, 4 °C, Tempshell
    Cooling frames, 4 °C, Tempshell
    Blue
    2...8 °C
    Log in for availability
    Log in for availability
    1 * 1 PAIRE
    1 * 1 PAIRE - 150,00 €
    150,00 €
    1 * 1 PAIRE
       150,00 €
  • DLTTA100036
    Cooling frames, 22 °C, Tempshell
    Cooling frames, 22 °C, Tempshell
    Green
    20...24 °C
    Log in for availability
    Log in for availability
    1 * 1 PAIRE
    1 * 1 PAIRE - 150,00 €
    150,00 €
    1 * 1 PAIRE
       150,00 €