Orders LinkContactUsLinkComponent
Micro pipette tips, OMIX®
Micro pipette tips, OMIX®
Supplier:   VARIAN
Micro pipette tips, OMIX®
Supplier:  VARIAN
Certificates
Micro pipette tips, OMIX®
Micro pipette tips, OMIX®
12 Options Available Below
Product Details & Documents
Bond Elut OMIX pipette tips reliably purify and enrich femtomole and picomole levels of peptides and proteins prior to MALDI-TOF or LC/MS/MS. The unique monolithic sorbent technology used in Bond Elut OMIX consistently outperforms other tips by delivering uniform flow and strong analyte-to-surface interactions. Bond Elut OMIX tips bind more peptides to deliver better sequence coverage.
  • Fast, uniform flow maximizes productivity and reproducibility
  • Minimal peptide losses lead to higher recoveries
  • Available in three phases and sizes to deliver better sequence coverage
Product Family Options
10 μl Mini-Bed
Product Information
  • Tip style
    Volume
    Packed
    Availability
    Price
  • OMIX Mini-Bed, C18
    0,5 - 2 µl
    1 rack, 96 each
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 376,00 €
    376,00 €
    1 * 1 ST
       376,00 €
  • OMIX Mini-Bed, C18
    0,5 - 2 µl
    6 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 576 ST
    1 * 576 ST - 1 690,00 €
    1 690,00 €
    1 * 576 ST
       1 690,00 €
10 μl
Product Information
  • Tip style
    Volume
    Packed
    Availability
    Price
  • OMIX, C4
    2 - 10 µl
    1 rack, 96 each
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST - 362,00 €
    362,00 €
    1 * 96 ST
       362,00 €
  • OMIX, C4
    2 - 10 µl
    6 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 576 ST
    1 * 576 ST - 1 350,00 €
    1 350,00 €
    1 * 576 ST
       1 350,00 €
  • OMIX, C18
    2 - 10 µl
    1 rack, 96 each
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 362,00 €
    362,00 €
    1 * 1 ST
       362,00 €
  • OMIX, C18
    2 - 10 µl
    6 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 576 ST
    1 * 576 ST - 1 660,00 €
    1 660,00 €
    1 * 576 ST
       1 660,00 €
100 μl
Product Information
  • Tip style
    Volume
    Packed
    Availability
    Price
  • OMIX SCX
    2 - 10 µl
    1×96 tips
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST - 369,00 €
    369,00 €
    1 * 96 ST
       369,00 €
  • OMIX C18
    10 - 100 µl
    1×96 tips
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 439,00 €
    439,00 €
    1 * 1 ST
       439,00 €
  • OMIX SCX
    10 - 100 µl
    1×96 tips
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST - 439,00 €
    439,00 €
    1 * 96 ST
       439,00 €
  • OMIX, C4
    10 - 100 µl
    6 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 576 ST
    1 * 576 ST - 1 830,00 €
    1 830,00 €
    1 * 576 ST
       1 830,00 €
  • OMIX, C4
    10 - 100 µl
    1 rack, 96 each
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST - 439,00 €
    439,00 €
    1 * 96 ST
       439,00 €
  • OMIX, C18
    10 - 100 µl
    6 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 576 ST
    1 * 576 ST - 2 100,00 €
    2 100,00 €
    1 * 576 ST
       2 100,00 €