Orders LinkContactUsLinkComponent
Floor cleaner, Quick Stripper, Ultan®
Floor cleaner, Quick Stripper, Ultan®
Quick Stripper, Ultan®, floor cleaner
Catalog # KAMM0404741
Supplier:   KAMMERER
Floor cleaner, Quick Stripper, Ultan®
Catalog # KAMM0404741
Supplier:  KAMMERER
Floor cleaner, Quick Stripper, Ultan®
Quick Stripper, Ultan®, floor cleaner
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents
High performance intensive cleaner, quickly and effectively removes polish residues and caked-on dirt.
  • Odour-free
  • Only a small amount required
  • Suitable for all alkali-resistant floor surfaces
Specifications
  • Description:
    Quick Stripper, Ultan®, floor cleaner