Orders LinkContactUsLinkComponent
Analytical HPLC columns, Microsorb™ Si
Analytical HPLC columns, Microsorb™ Si
Supplier:   VARIAN
Analytical HPLC columns, Microsorb™ Si
Supplier:  VARIAN
Certificates
Analytical HPLC columns, Microsorb™ Si
3 Options Available Below
Product Details & Documents
Microsorb™ Silica is ideal for the separation of low molecular weight compounds that dissolve in organic solvents. Separations on silica columns depend upon the difference in orientation, type and number of functional groups associated with the compounds in the sample.
Product Family Options
Product Information
  • Column Format
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Analytical
    100 Å
    3 µm
    4,60 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 871,00 €
    871,00 €
    1 * 1 ST
       871,00 €
  • Analytical
    100 Å
    5 µm
    4,60 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Analytical
    100 Å
    5 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 982,00 €
    982,00 €
    1 * 1 ST
       982,00 €