ContactUsLinkComponent
Seal roller, QuickSeal Hand Roller™
Seal roller, QuickSeal Hand Roller™
  731-0461
undefined
Seal roller, QuickSeal Hand Roller™
  731-0461
  • Description:
    Seal applicator, QuickSeal Hand Roller™
  • Pk:
    1
This adhesive seal roller ensures even pressure is applied across the seal for a complete application to the plate.
The handle of the roller is made of a durable plastic, with a semi-hard padded rolling wheel.